Would you like to log in again?
Synthware Outer Joint Adapters with Glass Stopcock, Kemtech America
Supplier: Kemtech America
Synthware
A691420 A692440
60000-260EA 70.08 USD InStock
60000-260 60000-264
Synthware Outer Joint Adapters with Glass Stopcock, Kemtech America
Adapters Adapters, Joints with Tube Connectors
Use this outer joint adapter with glass stopcock for regulating the input of gases or liquids into reaction systems.
  • Outer [ST] joint
  • Glass stopcock
  • With hose connection
Featuring a hose connection size of 8 mm O.D. Stopcock bore is 2 mm.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty