Would you like to log in again?
Synthware Flushing Adapters, Kemtech America
Supplier: Kemtech America
Synthware
A681922 A681420 A682440
60000-270EA 139.03 USD InStock
60000-270 60000-272 60000-268
Synthware Flushing Adapters, Kemtech America
Adapters Adapters, Joints with Tube Connectors
The flushing adapter is used to flush and fill attached vessels.
  • Inner [ST] joint
  • T-bore PTFE stopcock
  • With hose connection
Adapter features an inner [ST] joint and 2mm T-bore PTFE stopcock. Hose connection O.D. is 8mm.
Ordering information: For 2 mm fluropolymer resin T-bore stopcock plug, contact your Avantor representative.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty