Would you like to log in again?
Synthware Vacuum/Inert Gas Adapters, Kemtech America
Supplier: Kemtech America
Synthware
A881420G A882440G A882440
60000-284EA 128.35 USD InStock
60000-284 60000-278 60000-276
Synthware Vacuum/Inert Gas Adapters, Kemtech America
Adapters Adapters, Joints with Tube Connectors
Use this adapter for vacuum/inert gas applications.
  • [ST] joint
  • With T-bore glass stopcock or T-bore PTFE plug
Adapter features a 4 mm T-bore glass stopcock having constant pressure retention device or 2 mm T-bore 1:5 taper PTFE plug.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty