Would you like to log in again?
Synthware Flow Control Adapters with Metering Stopcock, 90° Angle, Kemtech America
Supplier: Kemtech America
Synthware
A722440 A721420 A721922
60000-294EA 121.07 USD InStock
60000-294 60000-296 60000-298
Synthware Flow Control Adapters with Metering Stopcock, 90° Angle, Kemtech America
Adapters Adapters, Joints with Tube Connectors
Provides on/off control with the main valve and allows delivery of preset volumes of gases or liquids to reactions using the needle valve.
  • 90° angle bend
  • [ST] joint
  • Metering stopcock
Stopcock bore is 2 mm.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty