Would you like to log in again?
Synthware Vacuum Adapters, Kemtech America
Supplier: Kemtech America
Synthware
A741420 A741420G A742440G A742440 A741922 A741922G
60000-350EA 84.03 USD InStock
60000-350 60000-344 60000-346 60000-352 60000-354 60000-342
Synthware Vacuum Adapters, Kemtech America
Adapters Adapters, Joints with Tube Connectors
Vacuum adapter is designed for evacuation or purging.
  • [ST] joint
  • PTFE or glass stopcock
Features a 2mm straight-bore PTFE or glass stopcock.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty