Would you like to log in again?
Synthware Transfer Valve Adapters, Kemtech America
Supplier: Kemtech America
Synthware
A392440 A391420
60000-558EA 172.7 USD InStock
60000-558 60000-562
Synthware Transfer Valve Adapters, Kemtech America
Adapters Adapters, Joints with Tube Connectors
Vacuum valves have two Viton® O-rings on the shaft and an integral PTFE O-ring on the tip.
  • Two Viton® O-rings on shaft
  • Integral PTFE O-ring on tip
Vacuum valve height is 0–4 mm. Vacuum plug/control knob 60004-014 has a PTFE O-ring at tip.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty