Would you like to log in again?
Synthware Distilling Adapters, 75°
Supplier: Kemtech America
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Synthware
A972440 A971922 A971420
60000-608EA 56.13 USD InStock
60000-608 60000-612 60000-610
Synthware Distilling Adapters, 75°
Distillation Components Distillation Adapters
Synthware distillation adapter with 75° angle.
  • 75° angle
  • Vacuum-tight seal
  • Accommodates thermometer
  • Compression cap and Viton® O-ring included
Features a compression cap and Viton® O-ring for a vacuum-tight seal of plain thermometer.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty