Would you like to log in again?
Synthware Inlet Adapters, Long Stem, Kemtech America
Supplier: Kemtech America
Synthware
A651420 A652440
60000-626EA 70.61 USD InStock
60000-626 60000-622
Synthware Inlet Adapters, Long Stem, Kemtech America
Adapters Adapters, Joints with Tube Connectors
These inlet adapters feature a long stem.
  • Top outer [ST] joint
  • Bottom inner [ST] joint
  • Sealed-in lower tube extending below inner joint
Adapters have top outer and lower inner [ST] joints and sealed-in lower tube extending below inner joint. Tube length is measured below joint.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty