Would you like to log in again?
Synthware Vacuum Distilling Adapter, Kemtech America
Supplier: Kemtech America
Synthware
A491420
60000-700EA 107.66 USD InStock
60000-700
Synthware Vacuum Distilling Adapter, Kemtech America
Distillation Components Distillation Adapters
This vacuum distillation adapter is useful in the sub-boiling separation of mixtures with volatile components.
  • 100mm span between joints
  • [ST] joints
The span between joints is 100 mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty