Would you like to log in again?
Synthware Gas Washing Bottle with Fritted Disc and Two Hose Connections, Kemtech America
Supplier: Kemtech America
Synthware
B300500C B300500X B300125X B300250C B300125C
60000-738EA 365.59 USD InStock
60000-738 60000-728 60000-736 60000-730 60000-732
Synthware Gas Washing Bottle with Fritted Disc and Two Hose Connections, Kemtech America
Bottles Gas Washing Bottles
Gas washing bottle with [ST]45/50 cap-style stopper and fritted disc angled at 15° for even distribution of gas throughout absorbing material.
  • Heavy wall borosilicate construction
  • Fritted disc
  • Large base
  • 45/50 ground glass joint
Available with a coarse or extra-coarse fritted disc. Springs (60000-758) are ordered separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty