Would you like to log in again?
Synthware Distilling Columns, Vigreux, Kemtech America
Supplier: Kemtech America
Synthware
C109130 C102443 C101100 C104150 C101420 C101922 C101075 C102441 C102442
60000-778EA 94.89 USD InStock
60000-778 60000-766 60000-768 60000-776 60000-764 60000-770 60000-780 60000-772 60000-782
Synthware Distilling Columns, Vigreux, Kemtech America
Distillation Components Distillation Columns
Column length is measured from the highest to lowest indent in the column.
  • Heavy wall borosilicate construction
  • Ground glass joints
  • Vigreux indentations
Vigreux column features various downward facing indentations that drastically increase surface area.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty