Would you like to log in again?
Synthware Low Form Dewar Flasks, Hemispherical, Kemtech America
Supplier: Kemtech America
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Synthware
F240100 F240250 F240500
60001-184EA 474.31 USD InStock
60001-184 60001-188 60001-186
Synthware Low Form Dewar Flasks, Hemispherical, Kemtech America
Dewars and Cryogenic Vessels
Flask size shown is the maximum capacity flask that will fit inside the dewar.
  • Heavy wall borosilicate inner construction
  • Aluminum housing
  • Weighted metal base
Hemispherical dewars work great with flasks up to the size listed.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty