Would you like to log in again?
Synthware Wide Mouth Dewar Flasks, Metal Housing, Kemtech America
Supplier: Kemtech America
Synthware
F111000 F114300 F110350 F110665
60001-204EA 948.48 USD InStock
60001-204 60001-198 60001-196 60001-200
Synthware Wide Mouth Dewar Flasks, Metal Housing, Kemtech America
Dewars and Cryogenic Vessels
Synthware Dewar flask with weighted metal base for stability and a full length aluminum housing which extends to the top rim for added protection.
  • Heavy wall borosilicate inner construction
  • Aluminum housing
  • Weighted metal base
Tall form dewards work well with cold traps for vacuum/pressur epumps.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty