Would you like to log in again?
Synthware Chromatography Flow Control Adapters, Kemtech America
Supplier: Kemtech America
Synthware
C1324 C1301
60001-206EA 144.22 USD InStock
60001-206 60001-210
Synthware Chromatography Flow Control Adapters, Kemtech America
Adapters Adapters, Joints with Tube Connectors
This adapter is used for chromatography flow control.
  • High vacuum valve
  • Metering tip
Adapter features a high vacuum valve (0-4 mm) with metering tip to adjust air pressure.
Ordering information: Available accessories include a vacuum metering plug/control knob, O-rings, clamps for ball joints, and metal clamps for [ST] joints. Contact your Avantor representative for more information.
Caution: Since the chromatography column is used with positive air pressure, it is highly recommended that the operator uses the column behind a safety shield.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty