Would you like to log in again?
Synthware Distillation Receiving Sets, Distribution Adapter, Kemtech America
Supplier: Kemtech America
Synthware
D201414 A552440 D201922 A551922 A551420 D202424
60002-126EA 58.13 USD InStock
60002-126 60002-128 60002-130 60002-132 60002-134 60002-136
Synthware Distillation Receiving Sets, Distribution Adapter, Kemtech America
Distillation Components Distillation Receivers
Rotating cow receiver allows four flasks to be progressively filled.
  • Heavy wall Borosilicate construction
  • Ground glass joint
  • Keck clips included
The distillation receiver set allows you to distill into three separate flasks.
Ordering information: Set components are sold separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty