Would you like to log in again?
Synthware Filtration Apparatus with 47 mm Stainless Steel Disc, [ST]40/35 Joint Connections, Kemtech America
Supplier: Kemtech America
Synthware
F10401LA
60002-276EA 614.03 USD InStock
60002-276
Synthware Filtration Apparatus with 47 mm Stainless Steel Disc, [ST]40/35 Joint Connections, Kemtech America
Filters Membrane Filter Units
Uses 47 mm membranes for filtering corrosive solutions, particulate removal from HPLC solvents, and filtration analysis with solid phase extraction disks.
  • Complete Kit
  • Heavy wall borosilicate construction
  • Ground glass joint
  • Stainless steel filter
Support base has removable 47 mm stainless steel support screen for use with viscous or proteinaceous solutions.
Delivery information: Complete assembly includes a 300 ml graduated funnel, aluminum clamp, support base, and 1 L filtering flask.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty