Would you like to log in again?
Synthware Recovery Flasks, Heavy-Wall, Kemtech America
Supplier: Kemtech America
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Synthware
F318250 F31291L F311950 F314250 F311910 F311425 F319100 F311200 F311250 F311405 F318500 F311450 F312425 F314100 F314200 F312450 F311410 F314500 F311925 F319250 F318100 F311100 F314000
60002-440EA 47.43 USD InStock
60002-440 60002-450 60002-420 60002-430 60002-452 60002-444 60002-422 60002-410 60002-454 60002-432 60002-424 60002-446 60002-412 60002-434 60002-456 60002-448 60002-426 60002-458 60002-414 60002-436 60002-428 60002-416 60002-418
Synthware Recovery Flasks, Heavy-Wall, Kemtech America
Flasks Distillation Flasks
Hand-blown from tubing to ensure uniform wall thickness.
  • Heavy wall borosilicate construction
  • Ground glass joint
  • Various sizes available
The Synthware recovery flask is designed for heating. Evaporation and storage or transfer of solvents.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty