Would you like to log in again?
Synthware Reaction Flasks with High Vacuum Valve
Supplier: Kemtech America
Synthware
F544250 F541250 F541950 F541100 F544100 F544500 F541050 F54241L F549250 F549100
60003-256EA 149.26 USD InStock
60003-256 60003-266 60003-264 60003-274 60003-262 60003-272 60003-260 60003-270 60003-258 60003-268
Synthware Reaction Flasks with High Vacuum Valve
Reaction Vessels Reaction Vessels, Flask Shape
Reaction flasks are equipped with a high vacuum valve that features a PTFE O-ring at the tip and a PTFE wiper to protect the Viton® O-ring on the shaft.
  • Heavy wall borosilicate construction
  • Ground glass joint
  • Teflon® covered high vacuum valve included
Valve size: 0 to 4 mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty