Would you like to log in again?
Synthware Gas Dispersion Tubes, Fritted, 90° Angle
Supplier: Kemtech America
Synthware
F284060C F283045C F282035M F284060M F284060X
60003-652EA 82.55 USD InStock
60003-652 60003-640 60003-650 60003-648 60003-644
Synthware Gas Dispersion Tubes, Fritted, 90° Angle
Gas Tubes
The 90° angle permits bubbles to be spread over a large area.
  • Heavy wall construction
  • 33 expansion borosilicate glass
  • Standard ground glass joints work with other brands
For use with filtration techniques.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty