Would you like to log in again?
Synthware Peptide Synthesis Vessels with T-Bore PTFE Stopcock and Vacuum, Solid Phase
Supplier: Kemtech America
Synthware
P140025M P140010M P140050M P140100M
60003-728EA 265.5 USD InStock
60003-728 60003-724 60003-732 60003-720
Synthware Peptide Synthesis Vessels with T-Bore PTFE Stopcock and Vacuum, Solid Phase
Reaction Vessels Reaction Vessels, Cylindrical Shape
Vessels feature an 8 mm O.D.
  • Teflon® Cap and stopcock
  • GL 14 Threaded cap
  • Medium fritted disc
  • 8 mm Sidearm and lower Ground glass joint
connection that is sealed on the inner joint for connection to an inner gas. Available with a fritted disc of coarse or medium porosity to support resin. The top of the vessel features a GL thread and a PTFE-lined screw cap. The PTFE stopcock has a 2 mm bore. The arm has an O.D. of 8 mm.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty