Would you like to log in again?
Synthware Photochemical Immersion Well without Joint, Quartz, Kemtech America
Supplier: Kemtech America
Synthware
V190390
60003-764EA 817.58 USD InStock
60003-764
Synthware Photochemical Immersion Well without Joint, Quartz, Kemtech America
Reaction Vessels Reaction Vessels, Photochemical
Double-walled quartz immersion well is designed for use with the photochemical reaction vessel with internal thread (see 60003-762 series) reactor and lamp.
  • Quartz construction
  • Double walled
A small diameter inner tube extends down the annular space to ensure coolant flows from the bottom of the well upward to the outlet. The well is equipped with an RC50 nylon bushing with Viton® O-ring to form a compression-type seal with threaded reaction vessels. The immersion well provides a greaseless connection and can be adjusted to a desired height. The absence of a ground joint allows for easy and quick dismantling.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty