Would you like to log in again?
Synthware Filter Funnel with Fritted Disc and Vacuum Valve, Kemtech America
Supplier: Kemtech America
Synthware
F371460M F374200M F374200C
60003-836EA 324.64 USD InStock
60003-836 60003-834 60003-832
Synthware Filter Funnel with Fritted Disc and Vacuum Valve, Kemtech America
Funnels Filtration Funnels Filter Funnels
Funnels are equipped with a 0 to 4 mm high vacuum valve and a 2 mm glass stopcock that functions as a constant pressure retention device.
  • Heavy wall borosilicate construction
  • Ground glass joints
  • Glass stopcock on top, Hi Vac Valve below
  • Coarse or medium fritted disc available
Available with coarse or medium porosity.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty