Would you like to log in again?
Synthware High Vacuum Valve, PTFE, O-Ring, T-Type
Supplier: Kemtech America
Synthware
S590012
60004-012EA 144.31 USD InStock
60004-012
Synthware High Vacuum Valve, PTFE, O-Ring, T-Type
Valves Vacuum Valves
With two 9 mm O.D. sidearms sealed to the barrel at a 90° angle to one another.
  • Heavy wall construction
  • 33 expansion Borosilicate glass
The non-rotating PTFE plug has three O-Rings. The tip O-Ring makes the primary seal on the precision formed taper seat. For replacement Viton O-Ring kits, see R110 and R112.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty