Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Synthware Valve, High Vacuum, PTFE-Protected, 180° Sidearms, Kemtech America

Supplier: Kemtech America

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Synthware
S620008 S620012 S620004
60004-030EA 135.81 USD InStock
60004-030 60004-026 60004-028
Synthware Valve, High Vacuum, PTFE-Protected, 180° Sidearms, Kemtech America
Valves Vacuum Valves
High vacuum valve with two 9 mm O.D.

  • Hi Vacuum valves
  • Heavy wall Borosilicate construction
  • Teflon® Protected

sidearms sealed to the barrel at a 180° angle. The Viton® O-rings on the PTFE plug shaft are protected with precision PTFE wipers. The tip of the plug has an integral O-ring to make the primary seal. This design provides an excellent vacuum-tight seal with PTFE as the only material exposed to chemicals.

Order Now
 

ORDER

ACCESSORIES

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty