Would you like to log in again?
Synthware Valve, High Vacuum, Metering, PTFE-Protected, Low Hold Up Sidearms
Supplier: Kemtech America
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Synthware
S680004 S680012 S680008
60004-066EA 153.51 USD InStock
60004-066 60004-062 60004-064
Synthware Valve, High Vacuum, Metering, PTFE-Protected, Low Hold Up Sidearms
Valves Vacuum Valves
High vacuum metering valve with two 9mm O.D
  • Hi vacuum valves
  • Heavy wall borosilicate construction
  • Teflon® protected
sidearms bent to minimize hold up of liquids. The Viton® O-rings on the PTFE plug shaft are protected with precision PTFE wipers. The tip of the plug has an integral O-ring to make the primary seal. This design provides an excellent vacuum-tight seal with PTFE as the only material exposed to chemicals.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty