Would you like to log in again?
Synthware Adapter, Thermometer, with Hose Connection, Microscale, Kemtech America
Supplier: Kemtech America
Synthware
A781410
60004-106EA 43.27 USD InStock
60004-106
Synthware Adapter, Thermometer, with Hose Connection, Microscale, Kemtech America
Adapters Adapters, Joints with Tube Connectors
Vacuum joint adapter with vertically adjustable condenser tube for microscale applications.
  • Vacuum-tight seal
  • Cap accommodates thermometer
  • Nylon compression cap and Viton® O-ring included
  • With hose connection
Supplied with a nylon compression cap and Viton® O-ring to form a vacuum tight seal. To use as a syringe port, replace the O-ring with PTFE -lined septum. Cap accommodates a 5.0 to 7.5 mm (ext. Ø) thermometer.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty