Would you like to log in again?
Synthware Conical Reaction Vials, Heavy Wall
Supplier: Kemtech America
Synthware
V131003 V130701 V130501 V131008 V131005 V130711
60004-164EA 17.68 USD InStock
60004-164 60004-166 60004-172 60004-162 60004-170 60004-168
Synthware Conical Reaction Vials, Heavy Wall
Vials
Graduated.[ST] Joint No.: 14/10.
  • Heavy wall Borosilicate construction
  • Threaded Ground glass joint
  • Graduated
  • Cap included
This heavy wall reaction vial is great for microscale reactions.
Ordering information: For replacement caps, Viton® O-rings, or septa, contact your Avantor representative.
Delivery information: Supplied with one compression cap, one Viton® O-ring, and one fluoropolymer resin-faced silicone rubber septum.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty