Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Synthware Vacuum/Inert Gas Manifolds, All Glass
Supplier: Kemtech America
Synthware
M401004 M411003 M431003 M421004 M451004 M441005 M421005 M401005 M451005 M181003 M431005 M421003 M181004 M181005 M411005 M431004 M411004
60004-448EA 801.37 USD InStock
60004-448 60004-436 60004-458 60004-438 60004-444 60004-454 60004-432 60004-468 60004-446 60004-434 60004-456 60004-440 60004-450 60004-464 60004-430 60004-452 60004-470
Synthware Vacuum/Inert Gas Manifolds, All Glass
Manifolds
All glass manifolds are equipped with 4 mm double oblique, high vacuum hollow glass stopcocks, which are precision ground with corresponding numbers on barrel and plug.
  • Hollow glass stopcocks (held in by vacuum)
  • Heavy wall borosilicate construction
  • 3, 4 or 5 port options available
Lower vacuum chambers on the stopcock are evacuated with a separate hole in the plug to secure the plug in the stopcock barrels. Beams have an ext. Ø of 22 mm, and are available in a variety of opening configurations. Distance between stopcocks is 100 mm center to center.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty