Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Synthware High Vacuum Manifold with Solid Glass Stopcock, Kemtech America
Supplier: Kemtech America
Synthware
M260003 M290004 M230005 M280004 M300005 M250003 M250004 M260005 M280005 M230003 M250005 M230004 M260004 M240004 M290005
60004-506EA 1273.72 USD InStock
60004-506 60004-504 60004-498 60004-476 60004-512 60004-500 60004-478 60004-472 60004-494 60004-486 60004-474 60004-480 60004-490 60004-482 60004-492
Synthware High Vacuum Manifold with Solid Glass Stopcock, Kemtech America
Manifolds
Manifolds are equipped with 4 mm double oblique, high vacuum solid glass stopcocks which are precision ground with corresponding numbers on barrel and plug.
  • Hollow glass stopcocks (held in by vacuum)
  • Heavy wall borosilicate construction
  • 3, 4 or 5 port options available
Solid glass stopcocks have constant pressure retention devices. Manifold beams have an ext. Ø of 22 mm and are available in a variety of opening configurations. Distance between stopcocks is 100 mm center to center.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty