Would you like to log in again?
Synthware Single Bank Vacuum Manifold with #15 O-Ring Joints, Kemtech America
Supplier: Kemtech America
Synthware
M311508
60004-594EA 743.05 USD InStock
60004-594
Synthware Single Bank Vacuum Manifold with #15 O-Ring Joints, Kemtech America
Manifolds
Manifold is equipped with four ports that are spaced 100 mm from center to center.
  • Hi vacuum valves
  • Heavy wall borosilicate construction
  • Four ports
  • #15 O-ring joints
The 0 to 8 mm high vacuum valves are mounted on a 45° angle to the operator for easy access. Overall width is 395 mm.
Ordering information: Accessory items must be ordered separately.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty