Would you like to log in again?
Synthware Vacuum Trap, Kemtech America
Supplier: Kemtech America
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Synthware
V104045 V103029 V103834 V105050 V105550 V105045
60004-708EA 178.36 USD InStock
60004-708 60004-704 60004-706 60004-712 60004-702 60004-710
Synthware Vacuum Trap, Kemtech America
Vacuum Traps
Traps feature [ST] joints for easy removal of condensate
  • Heavy wall borosilicate construction
  • 55/50 connecting joint
  • 10 mm center tube
Center tube connection has an O.D. of 10mm. Side tube hose connection is 13mm. Body length is measured below the joint.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty