Would you like to log in again?
Synthware Solvent Removal Vacuum Trap, Kemtech America
Supplier: Kemtech America
Synthware
V121000 V120500
60004-716EA 207.48 USD InStock
60004-716 60004-714
Synthware Solvent Removal Vacuum Trap, Kemtech America
Vacuum Traps
Trap is designed for large volume solvent removal by vacuum.
  • 45/50 joint connects two components
  • Heavy wall borosilicate construction
With [ST] 40/35 joint. Center tube connection has an ext. Ø of 12 mm. The side tube connection has an ext. Ø of 10 mm.
Ordering information: Trap 60004-714 fits a 1900 ml Dewar flask. Trap 60004-716 fits a 4300 ml Dewar flask.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty