Would you like to log in again?
Synthware Solvent Still Head, Kemtech America
Supplier: Kemtech America
Synthware
S234004 S234504
60004-756EA 457.5 USD InStock
60004-756 60004-754
Synthware Solvent Still Head, Kemtech America
Distillation Components Distillation Heads
Similar to repurification solvent still (see 60004-750 series), the solvent still head features a drip tip on the delivery port in addition to the [ST] 14/20 inner joint.
  • Heavy wall Borosilicate construction
  • Ground glass joint
  • Teflon® stopcocks
  • Extended drip tip
Supplied with a 4 mm straight bore stopcock in the upper portion for easy syringe needle access.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty