Would you like to log in again?
Synthware Syringe Needles, Stainless Steel, Kemtech America
Supplier: Kemtech America
Synthware
S389200 S386200 S386150 S380230 S380216 S389300 S389150 S380221 S386300
60004-792EA 13.38 USD InStock
60004-792 60004-794 76264-420 76264-134 76264-138 76264-136 76264-422 60004-796 76264-418
Synthware Syringe Needles, Stainless Steel, Kemtech America
Syringe Needles
Stainless steel needles are designed with extra length for solvent retrieval.
  • Stainless steel construction
  • Luer lock design
These long needles are ideal for solvent retrieval.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty