Would you like to log in again?
Synthware Solid Pennyhead [ST] Stoppers, Glass, Kemtech America
Supplier: Kemtech America
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Synthware
S291018 S292942 S291030 S291420 S291922 S292440
60004-818EA 16.87 USD InStock
60004-818 60004-826 60004-816 60004-822 60004-824 60004-820
Synthware Solid Pennyhead [ST] Stoppers, Glass, Kemtech America
Stoppers Glass Stoppers
Solid glass penny head stopper.
  • Heavy wall Borosilicate construction
  • Male ground glass joint
  • Penny head design
  • Solid
Penny head design allows for easy removal of the stopper.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty