Would you like to log in again?
Synthware PTFE Stoppers, Solid and Hollow
Supplier: Kemtech America
Synthware
S531018 S531922 S531420 S532440
60004-906EA 51.26 USD InStock
60004-906 60004-902 60004-904 60004-900
Synthware PTFE Stoppers, Solid and Hollow
Stoppers PE/LDPE/HDPE Stoppers
PTFE stopper features a nylon extraction nut that permits easy removal of the stopper from the glass outer joint.
  • Extraction nut
  • Teflon® construction
Available in solid or hollow formats.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty