Would you like to log in again?
Synthware 'T' Shaped Tubing Connectors, Kemtech America
Supplier: Kemtech America
T121000 T120800
60004-914EA 20.21 USD InStock
60004-914 60004-916
Synthware 'T' Shaped Tubing Connectors, Kemtech America
Fittings and Connectors
The "T" shaped tubing connectors are manufactured from Synthware glass.
  • 8 mm tube connector
  • Heay wall borosilicate glass
T-Shaped connector allows for an inline tubing addition.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty