Would you like to log in again?
Synthware Four Finger Clamp, Kemtech America
Supplier: Kemtech America
Synthware
W170004
60005-030EA 28.24 USD InStock
60005-030
Synthware Four Finger Clamp, Kemtech America
Clamps General Purpose Clamps
Four finger clamp is constructed of die-cast alloy, with a nickel/chrome plated finish.
  • Cork lined fingers
  • Adjustable
  • Chromed plated
  • Plastic wing nut style tension screw
Clamps feature cork lined interlocking jaws. They accept articles up to 80 mm in diameter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty