Would you like to log in again?
Synthware Three Finger Double Adjustable Clamp, Kemtech America
Supplier: Kemtech America
Synthware
W192203 W181103
60005-034EA 26.13 USD InStock
60005-034 60005-032
Synthware Three Finger Double Adjustable Clamp, Kemtech America
Clamps General Purpose Clamps
Double adjustable clamp with PVC coated prongs.
  • Adjustable
  • Plastic coated fingers
  • Round plastic tension screws
  • Chrome plated
Clamps are constructed of chrome-plated Zinc-alloy. Small clamp accepts objects from 0 to 60 mm. Large clamp accepts object from 0 to 102 mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty