Would you like to log in again?
Synthware Stopper, Cap Design, Kemtech America
Supplier: Kemtech America
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Synthware
S391420 S392942 S392440 S391656
60004-848EA 25.86 USD InStock
60004-848 60005-196 60004-844 60004-850
Synthware Stopper, Cap Design, Kemtech America
Stoppers Glass Stoppers
Designed for use with an apparatus equipped with an inner opening.
  • Heavy wall borosilicate construction
  • Solid glass plug
  • Double oblique
  • Retention clip lncluded Female Ground glass joint
For apparatus having an inner opening.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty