Would you like to log in again?
Synthware High Vacuum Valves, PTFE, O-Ring, 180°
Supplier: Kemtech America
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Synthware
S560004 S560008
60005-236EA 134.76 USD InStock
60005-236 60005-234
Synthware High Vacuum Valves, PTFE, O-Ring, 180°
Valves Vacuum Valves
The non-rotating PTFE shaft has three O-Rings. The tip O-Ring makes the primary seal on the precision formed taper seat.
  • Heavy wall construction
  • 33 expansion Borosilicate glass
For replacement Viton O-Ring kits, see R007, R010, or R108 and R110.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty