Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Talon® Small Lab-Frames
Supplier: VWR
VWR® Talon®
80862683 80862677
60075-008EA 702.96 USD InStock
60075-008 60079-552
VWR® Talon® Small Lab-Frames
Support Rods and Frames
Great for glassware setups in small laboratories or where space is limited.
  • For vertical mounting
  • Ideal for limited-space glassware setups
  • Permanently mountable base
Frames measure 610Lx610Hmm (24x24") with a 457mm (18") wide base for stability. Base may be permanently mounted to benchtop with screws (included).
Ordering information: Each Lab Frame includes eight 51 mm (2") rods, two 457 mm (18") rods, eight 610 mm (24") rods, four rod end connectors, eighteen S-connectors, and four Lab Frame feet.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty