Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Talon® Medium Lab-Frames
Supplier: VWR
VWR® Talon®
80862684 80862678
60079-554EA 1244.7 USD InStock
60079-554 60075-009
VWR® Talon® Medium Lab-Frames
Support Rods and Frames
Lab Frames are convenient for distillation and general setups.
  • For horizontal or vertical mounting
  • Convenient size for distillation and general setups
  • Stable base
Can be assembled as 1220Lx610Hmm (48x24") horizontal or 610Lx1220Hmm (24x48") vertical frame. Base is 46 cm (18") wide for stability.
Ordering information: Each Lab Frame includes eight 51 mm (2") rods, two 457 mm (18") rods, seven 610 mm (24") rods, four 1220 mm (48") rods, four rod end connectors, thirty-five S-connectors, and four Lab Frame feet.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty