Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Talon® Large Lab-Frames
Supplier: VWR
VWR® Talon®
80862679 80862685
60079-556EA 1967.88 USD InStock
60079-556 60077-003
VWR® Talon® Large Lab-Frames
Support Rods and Frames
These 1220x1220mm (48x48") frames are ideal for complex glassware setups.
  • For large-capacity setups
  • Stable
  • Ideal for complex glass setups
Base is 460mm (18") wide for stability.
Ordering information: Each Lab Frame includes ten 51 mm (2") rods, three 457 mm (18") rods, two 914 mm (36") rods, ten 1220 mm (48") rods, four rod end connectors, thirty-eight S-connectors, and six Lab Frame feet.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty