Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Talon® Extra-Large Lab-Frames
Supplier: VWR
VWR® Talon®
80862686 80862680
60079-558EA 2964.03 USD InStock
60079-558 60078-006
VWR® Talon® Extra-Large Lab-Frames
Support Rods and Frames
Extra-large lab frames offer the greatest versatility for your most demanding setups.
  • Extra large design
  • For horizontal or vertical use
  • Versatile and adaptable
  • Mountable on either side
Measure 1220Lx1830Hmm (48x72"). May be used either horizontally or vertically. Include six 305mm (12") rods and feet for securing frame to wall. Apparatus mountable on either side of frame.
Ordering information: Each Lab Frame includes six 51 mm (2") rods, six 305 mm (12") rods, two 610 mm (24") rods, seven 1220 mm (48") rods, five 1829 mm (72") rods, four rod end connectors, forty-five S-connectors, and eight Lab Frame feet.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty