Would you like to log in again?
VWR® Talon® Aluminum Support Plate
Supplier: VWR
VWR® Talon®
80862634
60079-000EA 113.55 USD InStock
60079-000
VWR® Talon® Aluminum Support Plate
Support Stands
Talon® Aluminum Support Plate fits standard lattice lab frames using VWR® Talon® connectors 21572-501, 21572-556, or 60097-055.
  • Aluminum construction
  • Compatible with standard lattice lab frames
  • Hold a variety of media vessels
Supports beakers, mantles, and Petri dishes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty