Would you like to log in again?
VWR® Talon® Multiple Rod Connector
Supplier: VWR
VWR® Talon®
80862643
60079-053CS 981.86 USD InStock
60079-053
VWR® Talon® Multiple Rod Connector
Support Rods and Frames
Multiple rod connector holds lab frame rods or extension clamps at 90° while allowing both horizontal and vertical adjustment of lattice after construction.
  • Versatile
  • Fully adjustable horizontally and vertically
  • Side and front holes
Versatility and maximum adjustment capability at your fingertips. Two adjacent rods (sold separately) fit into oval hole in connector that prevent turning when tightened. Create a variety of configurations via side and front holes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty