Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Talon® Horizontal Mounting Bars with Coupler
Supplier: VWR
VWR® Talon®
80862648 80862646 80862647
60079-104CS 180.8 USD InStock
60079-104 60079-107 60079-106
VWR® Talon® Horizontal Mounting Bars with Coupler
Support Rods and Frames
Horizontal mounting bars feature a threaded end for direct installation into channels.
  • For use with steel channel frames
  • Locking nut included
  • Provided with channel coupler
Supplied with spring coupler for use in 41mm (119/32") steel channels.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty