Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Talon® Rods, Aluminum
Supplier: VWR
VWR® Talon®
80862652 80862659 80862657 80862654 80862655 80862656 80862658 80862660 80862653 80862661 80862650
60079-406CS 300.42 USD InStock
60079-406 60079-428 60079-381 60079-512 60079-325 60079-348 60079-508 60079-360 60079-440 60079-483 60079-461
VWR® Talon® Rods, Aluminum
Support Rods and Frames
These hard aluminum rods provide smooth fit with lab frames.
  • For use with VWR® Talon® lab frames
  • Resistant to corrosion
  • Precise fit
  • Chamfered ends
Center-less ground for precise fit. Corrosion-resistant with chamfered ends for easy location.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty