Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Talon® Rods, Stainless Steel
Supplier: VWR
VWR® Talon®
80862662 80862669 80862666 80862663 80862670 80862664 80862668 80862665 80862667 80862671 80862672
60079-538CS 1072.25 USD InStock
60079-538 60079-537 60079-539 60079-534 60079-533 60079-536 60079-535 60079-530 60079-540 60079-532 60079-531
VWR® Talon® Rods, Stainless Steel
Support Rods and Frames
Choose VWR® top-of-the-line rods when you need maximum strength and durability.
  • Strong and resilient
  • For use with VWR® Talon® lab frames
  • Stainless steel construction
Constructed of Type 303 stainless steel.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty